Cadmium acetate dihydrate, 99.999% (metals basis)
Cadmium acetate dihydrate, 99.999% (metals basis)
Cadmium acetate dihydrate, 99.999% (metals basis)
Thermo Scientific Chemicals

Cadmium acetate dihydrate, 99.999% (metals basis)

CAS: 5743-04-4 | C4H10CdO6 | 266.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011865.22
also known as 011865-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS5743-04-4
IUPAC Namecadmium(2+) diacetate dihydrate
Molecular FormulaC4H10CdO6
InChI KeyAUIZLSZEDUYGDE-UHFFFAOYSA-L
SMILESO.O.[Cd++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
It is used in dyestuff intermediates, ceramics and producing iridescent effects on porcelains and pottery. It acts as a reagent for determination of S, Se, and Te. It is also used in cadmium electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only