Sodium hexachloroiridate(IV) hexahydrate, Ir 33.9% min
Sodium hexachloroiridate(IV) hexahydrate, Ir 33.9% min
Sodium hexachloroiridate(IV) hexahydrate, Ir 33.9% min
Thermo Scientific Chemicals

Sodium hexachloroiridate(IV) hexahydrate, Ir 33.9% min

CAS: 19567-78-3 | Cl6H12IrNa2O6 | 558.987 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 011890.06
also known as 011890-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS19567-78-3
IUPAC Namedisodium hexachloroiridiumdiuide hexahydrate
Molecular FormulaCl6H12IrNa2O6
InChI KeyQZVJPVJIAOOTJU-UHFFFAOYSA-H
SMILESO.O.O.O.O.O.[Na+].[Na+].Cl[Ir--](Cl)(Cl)(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ir 33.9% min
Sodium hexachloroiridate(IV) hexahydrate is used pharmaceutical Intermediates and organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only