Gold rod, 3mm (0.1in) dia, Premion™, 99.9985% (metals basis)
Gold rod, 3mm (0.1in) dia, Premion™, 99.9985% (metals basis)
Gold rod, 3mm (0.1in) dia, Premion™, 99.9985% (metals basis)
Thermo Scientific Chemicals

Gold rod, 3mm (0.1in) dia, Premion™, 99.9985% (metals basis)

CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Catalog NumberQuantity
011892.06
also known as 011892-06
5 g
Catalog number 011892.06
also known as 011892-06
Price (USD)
1,458.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
1,458.00
Each
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
FormRod, 3mm (0.1in) diameter.
Assay from Supplier's CofA≥99.9985% (metals basis)
Length≈3.7cm
Appearance (Color)Gold
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only