Tin wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis)
Tin wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis)
Tin wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis)
Thermo Scientific Chemicals

Tin wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis)

Form: approx. 5.72g/m | CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 cm
250 cm
1000 cm
Catalog number 011905.BW
also known as 011905-BW
Price (USD)
40.50
Each
Quantity:
50 cm
Request bulk or custom format
Price (USD)
40.50
Each
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only