Thallium granules, 6mm (0.2in) & down, 99.99% (metals basis), packaged in water
Thallium granules, 6mm (0.2in) & down, 99.99% (metals basis), packaged in water
Thallium granules, 6mm (0.2in) & down, 99.99% (metals basis), packaged in water
Thermo Scientific Chemicals

Thallium granules, 6mm (0.2in) & down, 99.99% (metals basis), packaged in water

CAS: 7440-28-0 | Tl | 204.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 012131.22
also known as 012131-22
Price (USD)
901.65
Online Exclusive
1,002.00
Save 100.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
901.65
Online Exclusive
1,002.00
Save 100.35 (10%)
Each
Chemical Identifiers
CAS7440-28-0
IUPAC Namethallium
Molecular FormulaTl
InChI KeyBKVIYDNLLOSFOA-UHFFFAOYSA-N
SMILES[Tl]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged in water
RUO – Research Use Only