Niobium carbide, 99+% (metals basis)
Niobium carbide, 99+% (metals basis)
Niobium carbide, 99+% (metals basis)
Thermo Scientific Chemicals

Niobium carbide, 99+% (metals basis)

CAS: 12069-94-2 | CNb | 104.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 012147.14
also known as 012147-14
Price (USD)
50.65
Special offer
Online exclusive
Ends: 28-Dec-2026
58.90
Save 8.25
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
50.65
Special offer
Online exclusive
Ends: 28-Dec-2026
58.90
Save 8.25
Each
Chemical Identifiers
CAS12069-94-2
IUPAC Namemethanidylidyneniobiumylium
Molecular FormulaCNb
InChI KeyAADPBXLVMHIYCT-UHFFFAOYSA-N
SMILES[C-]#[Nb+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.9% max.
Niobium carbide is used in tool bits for cutting tools.  Zirconium carbide and niobium carbide can be used as refractory coatings in nuclear reactors. It is used to produce arc-welding rods and in corrosion-resistant steel. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only