Sodium hexachloroosmate(IV) dihydrate, Os 38.7% min
Sodium hexachloroosmate(IV) dihydrate, Os 38.7% min
Sodium hexachloroosmate(IV) dihydrate, Os 38.7% min
Thermo Scientific Chemicals

Sodium hexachloroosmate(IV) dihydrate, Os 38.7% min

CAS: 1307-81-9 | Cl6Na2Os | 448.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
5 g
Catalog number 012176.06
also known as 012176-06
Price (USD)
666.65
Online Exclusive
741.00
Save 74.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
666.65
Online Exclusive
741.00
Save 74.35 (10%)
Each
Chemical Identifiers
CAS1307-81-9
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Os 38.7% min
Sodium hexachloroosmate(IV) dihydrate is used as an intermediate and catalyst in organic synthesis. It is also used to prepare dihydrogen complexes of osmium(II) containing meso-tetraphos-1 ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only