Potassium hexachloroosmate(IV), Os 38.7% min
Potassium hexachloroosmate(IV), Os 38.7% min
Potassium hexachloroosmate(IV), Os 38.7% min
Thermo Scientific Chemicals

Potassium hexachloroosmate(IV), Os 38.7% min

CAS: 16871-60-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
5 g
Catalog number 012177.03
also known as 012177-03
Price (USD)
230.65
Online Exclusive
256.00
Save 25.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
230.65
Online Exclusive
256.00
Save 25.35 (10%)
Each
Chemical Identifiers
CAS16871-60-6
IUPAC Nameosmium(4+) dipotassium hexachloride
Molecular FormulaCl6K2Os
InChI KeyVGKQJCSDERXWRV-UHFFFAOYSA-H
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[K+].[K+].[Os+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Os 38.7% min
Potassium hexachloroosmate(IV) used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only