Molybdenum carbide, 99.5% (metals basis)
Molybdenum carbide, 99.5% (metals basis)
Molybdenum carbide, 99.5% (metals basis)
Molybdenum carbide, 99.5% (metals basis)
Thermo Scientific Chemicals

Molybdenum carbide, 99.5% (metals basis)

CAS: 12069-89-5 | CMo2 | 203.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 012192.36
also known as 012192-36
Price (USD)
413.65
Special offer
Online exclusive
Ends: 30-Jun-2026
486.00
Save 72.35 (15%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
413.65
Special offer
Online exclusive
Ends: 30-Jun-2026
486.00
Save 72.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS12069-89-5
IUPAC Name(molybdeniomethylidene)molybdenum
Molecular FormulaCMo2
InChI KeyQIJNJJZPYXGIQM-UHFFFAOYSA-N
SMILES[Mo]=C=[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities≤0.5%
Appearance (Color)Black or grey to dark grey
Assay (unspecified)5.5-5.9% C
Molybdenum carbide is used in the production of solid molybdenum carbide grades of hard metal in the manufacture of mixed crystal. It used as coating material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only