Thermo Scientific Chemicals

Barium nitrate, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 012194.36
500 g, Each
Thermo Scientific Chemicals

Barium nitrate, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 012194.36
500 g, Each
Quantity
Catalog number: 012194.36
also known as 012194-36
Price (USD)
Quantity
-

Chemical Identifiers

CAS
10022-31-8
IUPAC Name
barium(2+) dinitrate
Molecular Formula
BaN2O6
InChI Key
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
SMILES
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Heavy Metals
(ICP-OMS) 5ppm
Impurity content
Sr 0.1%
pH
of a 5% solution 5.0-8.0 at 25?
Assay (unspecified)
99.0% Ba(NO₃)₂ min
Impurity content
Cl 5ppm

Description

Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text