Strontium hydroxide octahydrate, tech.
Strontium hydroxide octahydrate, tech.
Strontium hydroxide octahydrate, tech.
Thermo Scientific Chemicals

Strontium hydroxide octahydrate, tech.

CAS: 1311-10-0 | H18O10Sr | 265.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2 kg
10 kg
Catalog number 012199.18
also known as 012199-18
Price (USD)
30.80
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
30.80
Each
Chemical Identifiers
CAS1311-10-0
IUPAC Namestrontium(2+) octahydrate dihydroxide
Molecular FormulaH18O10Sr
InChI KeyUJPWWRPNIRRCPJ-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.[OH-].[OH-].[Sr++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)94% min
Strontium hydroxide octahydrate is used in the sugar industry for its purification, oil and paint industry to improve its drying properties and as an active ingredient in depilatory hair removal products. It is also used as a raw material for manufacturing of other products of strontium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only