Strontium fluoride, 99% min
Strontium fluoride, 99% min
Strontium fluoride, 99% min
Thermo Scientific Chemicals

Strontium fluoride, 99% min

CAS: 7783-48-4 | F2Sr | 125.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 012201.A1
also known as 012201-A1
Price (USD)
141.65
Online Exclusive
157.00
Save 15.35 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
141.65
Online Exclusive
157.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS7783-48-4
IUPAC Namestrontium(2+) difluoride
Molecular FormulaF2Sr
InChI KeyFVRNDBHWWSPNOM-UHFFFAOYSA-L
SMILES[F-].[F-].[Sr++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99% min
Strontium fluoride is used as an optical material for a small range of special applications. It also used as use is as a carrier of strontium-90 radioisotope in radioisotope thermoelectric generators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only