Strontium acetate hemihydrate, Reagent Grade
Strontium acetate hemihydrate, Reagent Grade
Strontium acetate hemihydrate, Reagent Grade
Thermo Scientific Chemicals

Strontium acetate hemihydrate, Reagent Grade

CAS: 14692-29-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 012203.30
also known as 012203-30
Price (USD)
68.65
Special offer
Online exclusive
Ends: 30-Jun-2026
80.90
Save 12.25 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
68.65
Special offer
Online exclusive
Ends: 30-Jun-2026
80.90
Save 12.25 (15%)
Each
Chemical Identifiers
CAS14692-29-6
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99% min
Strontium acetate is used as a catalyst, as a chemical intermediate and in medicine. The primary application of strontium acetate is as an active component in toothpastes. This compound is used to desensitize teeth to extremes of hot and cold, sweets, acids, or touch.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only