Indium foil, 0.127mm (0.005in) thick, 99.99% (metals basis)
Indium foil, 0.127mm (0.005in) thick, 99.99% (metals basis)
Indium foil, 0.127mm (0.005in) thick, 99.99% (metals basis)
Indium foil, 0.127mm (0.005in) thick, 99.99% (metals basis)
Thermo Scientific Chemicals

Indium foil, 0.127mm (0.005in) thick, 99.99% (metals basis)

Form: approx. 4.6g/50x50mm | CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
50 x 100 mm
100 x 150 mm
150 x 300 mm
Catalog number 012206.FI
also known as 012206-FI
Price (USD)
94.00
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
94.00
Each
Chemical Identifiers
CAS7440-74-6
IUPAC Nameindium
Molecular FormulaIn
InChI KeyAPFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver to silver-grey
FormFoil
Assay (unspecified)Indium (In): ≥99.99% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only