Platinum Rhodium gauze, 80 mesh woven from 0.076mm (0.003in) dia wire, 99.9% (metals basis)
Thermo Scientific Chemicals

Platinum Rhodium gauze, 80 mesh woven from 0.076mm (0.003in) dia wire, 99.9% (metals basis)

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 012217.FF
also known as 012217-FF
Price (USD)
-
Quantity:
25 x 25 mm
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
Avg. Mol. Wt. or Mol. Wt. RangePt:Rh; 90:10 wt%
Chemical Name or MaterialPlatinum Rhodium gauze
MDL NumberMFCD00198186
Mesh Size80 Mesh
View more
Platinum-rhodium alloy gauze serves as a catalyst in the oxidation of the ammonia from the Haber Process by atmospheric oxygen to produce nitric oxide and subsequently nitric acid, which is used for producing nitrogen fertilizers, ammonium nitrate, adipic acid, and toluene diisocyanate (precursor for polyurethane). It has high degree of chemical inertness and stability at very high temperatures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only