Lead(II) oxide, 99.9%
Lead(II) oxide, 99.9%
Lead(II) oxide, 99.9%
Lead(II) oxide, 99.9%
Thermo Scientific Chemicals

Lead(II) oxide, 99.9%

CAS: 1317-36-8 | OPb | 223.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012220.36
also known as 012220-36
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS1317-36-8
IUPAC Nameplumbanone
Molecular FormulaOPb
InChI KeyYEXPOXQUZXUXJW-UHFFFAOYSA-N
SMILESO=[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥99.90% (unspecified)
Appearance (Color)Pale yellow to yellow or pale orange
FormPowder or crystalline powder or crystals
It is employed mostly in lead-based industrial glass and industrial ceramics, including computer components. Lead monoxide is used as an intermediate/precursor in the manufacture of several products, for example water proof cements, lubricants, lubricating oils, inorganic pigments, lead soaps, petroleum refining, rubber, cathode ray tube glass, and polyvinyl chloride (PVC). It is useful for lead acid batteries as cathode and anode. Lead monoxide scores significant applications in oil, gas and chemical manufactures. It is an efficient catalyst for condensation reactions in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only