Cobalt(II) acetate tetrahydrate, Co 24%
Cobalt(II) acetate tetrahydrate, Co 24%
Cobalt(II) acetate tetrahydrate, Co 24%
Thermo Scientific Chemicals

Cobalt(II) acetate tetrahydrate, Co 24%

CAS: 6147-53-1 | C4H14CoO8 | 249.081 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 012227.30
also known as 012227-30
Price (USD)
59.65
Online exclusive
66.30
Save 6.65
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
59.65
Online exclusive
66.30
Save 6.65
Each
Chemical Identifiers
CAS6147-53-1
IUPAC Nameλ²-cobalt(2+) diacetate tetrahydrate
Molecular FormulaC4H14CoO8
InChI KeyZBYYWKJVSFHYJL-UHFFFAOYSA-L
SMILESO.O.O.O.[Co++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofACobalt, Co: ≥23.5%
FormCrystals or powder or crystalline powder
Appearance (Color)Red
It is used as an industrial catalyst that allow paints and varnishes to harden. Also as a precursor to various oil drying agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only