Cadmium nitrate tetrahydrate, 98.5% min
Cadmium nitrate tetrahydrate, 98.5% min
Cadmium nitrate tetrahydrate, 98.5% min
Thermo Scientific Chemicals

Cadmium nitrate tetrahydrate, 98.5% min

CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012228.22
also known as 012228-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS10022-68-1
IUPAC Namecadmium(2+) bis(nitric acid)
Molecular FormulaCdH2N2O6
InChI KeyIZIPRBSCGMISKX-UHFFFAOYSA-N
SMILES[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.5% min
Cadmium nitrate is used in manufacturing of cadmium salts, photographic emulsions, laboratory reagent and coloring glass. It is used for porcelain and as a flash powder in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only