Bismuth(III) nitrate pentahydrate, ACS, 98% min
Bismuth(III) nitrate pentahydrate, ACS, 98% min
Bismuth(III) nitrate pentahydrate, ACS, 98% min
Thermo Scientific Chemicals

Bismuth(III) nitrate pentahydrate, ACS, 98% min

CAS: 10035-06-0 | BiH10N3O14 | 485.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 012231.30
also known as 012231-30
Price (USD)
56.65
Special offer
Online exclusive
Ends: 28-Dec-2026
67.00
Save 10.35
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
56.65
Special offer
Online exclusive
Ends: 28-Dec-2026
67.00
Save 10.35
Each
Chemical Identifiers
CAS10035-06-0
IUPAC Namebismuth(3+) pentahydrate trinitrate
Molecular FormulaBiH10N3O14
InChI KeyFBXVOTBTGXARNA-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0% Bi(NO₃)₃?5H₂O min
Impurity contentAs 0.001%
Impurity contentCl 0.001%
Impurity contentCu 0.002%
Impurity contentFe 0.001%
View more
Luminous paints, preparation of bismuth saltsBismuth(III) nitrate pentahydrate is an oxidant for a variety of 4-substituted Hantzsch 1,4-dihydropyridines. It also serves as a reagent for selective oxidation of sufides to sulfoxides. Further, it is involved in the preparation of Dragendorff reagent and used as a TLC stain. It is also used to prepare other bismuth compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only