Tin(IV) oxide, 99.9% (metals basis)
Tin(IV) oxide, 99.9% (metals basis)
Tin(IV) oxide, 99.9% (metals basis)
Tin(IV) oxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Tin(IV) oxide, 99.9% (metals basis)

CAS: 18282-10-5 | O2Sn | 150.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 012283.22
also known as 012283-22
Price (USD)
57.10
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
57.10
Each
Chemical Identifiers
CAS18282-10-5
IUPAC Namestannanedione
Molecular FormulaO2Sn
InChI KeyXOLBLPGZBRYERU-UHFFFAOYSA-N
SMILESO=[Sn]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
It is employed in the manufacture of white enamels and for polishing glass. It has a broad range of applications in many areas such as semiconductors, photovoltaic materials, solar cells, lithium ion batteries, photocatalysts, and as gas-sensing agents in solid-state detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only