Mercury(II) iodide, ACS, 99.0% min (Assay-dried basis)
Mercury(II) iodide, ACS, 99.0% min (Assay-dried basis)
Mercury(II) iodide, ACS, 99.0% min (Assay-dried basis)
Thermo Scientific Chemicals

Mercury(II) iodide, ACS, 99.0% min (Assay-dried basis)

For Use As An Analytical Reagent For the Preparation of Nessler'S Reagent. | CAS: 7774-29-0 | HgI2 | 454.401 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012289.22
also known as 012289-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7774-29-0
IUPAC Namemercury(2+) diiodide
Molecular FormulaHgI2
InChI KeyYFDLHELOZYVNJE-UHFFFAOYSA-L
SMILES[I-].[I-].[Hg++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0% HgI₂ min
CommentSoluble mercury salts (as Hg) 0.05%
Impurity contentMercurous mercury (as Hg) 0.1%
Solution Testin potassium iodide solution P.T.
As an analytical reagent for the preparation of Nessler's Reagent. It is also used as an oxidizing agent for the synthesis of quinoxaline. Polycrystalline films of mercury (II) iodide, also play an important role in X-ray imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only