Calcium chloride dihydrate, 99% min
Calcium chloride dihydrate, 99% min
Calcium chloride dihydrate, 99% min
Calcium chloride dihydrate, 99% min
Thermo Scientific Chemicals

Calcium chloride dihydrate, 99% min

CAS: 10035-04-8 | CaCl2·2H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 012312.36
also known as 012312-36
Price (USD)
73.65
Special offer
Online exclusive
Ends: 28-Dec-2026
86.00
Save 12.35
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
73.65
Special offer
Online exclusive
Ends: 28-Dec-2026
86.00
Save 12.35
Each
Chemical Identifiers
CAS10035-04-8
IUPAC Namecalcium dihydrate dichloride
Molecular FormulaCaCl2H4O2
InChI KeyLLSDKQJKOVVTOJ-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (special)99.0-107.0%
FormGranular
Calcium chloride dihydrate is widely used as an ingredient in ceramic slipware, cosmetic and food items. It acts as a pH buffer in swimming pool water. It provides inhibition of swelling clays in the water phase of invert emulsion drilling fluids. It is employed in the oil industry to increase the density of solids-free brines. It is used as a flux in the Davy process for the manufacture of sodium through the electrolysis of molten sodium chloride. It finds an important application as an additive in plastics, fire extinguishers, blast furnaces and in wastewater treatment as a drainage aid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only