Tungsten(VI) chloride, 99%
Tungsten(VI) chloride, 99%
Tungsten(VI) chloride, 99%
Thermo Scientific Chemicals

Tungsten(VI) chloride, 99%

CAS: 13283-01-7 | Cl6W | 396.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
250 g
Catalog number 012337.09
also known as 012337-09
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
71.00
Save 10.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
71.00
Save 10.35 (15%)
Each
Chemical Identifiers
CAS13283-01-7
IUPAC Nametungsten hexachloride
Molecular FormulaCl6H6W
InChI KeyXHYHOBJDYHRWHO-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.Cl.Cl.[W]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
It is a precursor for many tungsten compounds. It is used in tungsten plating, purification of tungsten, and olefin polymerization. It is also used as petroleum catalyst, and in intelligent glass. It can be used for deoxygenation of epoxides and as a reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only