Manganese(II) acetate tetrahydrate, Mn 22% (typical)
Manganese(II) acetate tetrahydrate, Mn 22% (typical)
Manganese(II) acetate tetrahydrate, Mn 22% (typical)
Thermo Scientific Chemicals

Manganese(II) acetate tetrahydrate, Mn 22% (typical)

CAS: 6156-78-1 | C4H14MnO8 | 245.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 012351.A3
also known as 012351-A3
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Quantity:
2 kg
Request bulk or custom format
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS6156-78-1
IUPAC Namemanganese(2+) diacetate tetrahydrate
Molecular FormulaC4H14MnO8
InChI KeyCESXSDZNZGSWSP-UHFFFAOYSA-L
SMILESO.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale pink to pink to pale red
FormCrystals or crystalline powder or lumps
Assay from Suppliers CofA≥22.0%
It is used as a mordant in dyeing, drier for paints and varnishes, an oxidation catalyst, and in the manufacture of bister. It is also used in the manufacture of fertilizer. It is used for the production of high purity manganese dioxide, manganese oxide hydrate, oxide-based catalysts, and catalysts for oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Mordant in dyeing, in the manufacture of bister, as a drier for paints and varnishes, and as an oxidation catalyst
RUO – Research Use Only