Rhodium black, 99.9% (metals basis)
Rhodium black, 99.9% (metals basis)
Rhodium black, 99.9% (metals basis)
Thermo Scientific Chemicals

Rhodium black, 99.9% (metals basis)

CAS: 7440-16-6 | Rh | 102.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 012353.02
also known as 012353-02
Price (USD)
-
Quantity:
0.5 g
Request bulk or custom format
Chemical Identifiers
CAS7440-16-6
IUPAC Namerhodium
Molecular FormulaRh
InChI KeyMHOVAHRLVXNVSD-UHFFFAOYSA-N
SMILES[Rh]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only