Ruthenium black, 99.9% (metals basis)
Ruthenium black, 99.9% (metals basis)
Ruthenium black, 99.9% (metals basis)
Thermo Scientific Chemicals

Ruthenium black, 99.9% (metals basis)

CAS: 7440-18-8 | Ru | 101.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 012354.09
also known as 012354-09
Price (USD)
1,108.65
Online Exclusive
1,232.00
Save 123.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
1,108.65
Online Exclusive
1,232.00
Save 123.35 (10%)
Each
Chemical Identifiers
CAS7440-18-8
IUPAC Nameruthenium
Molecular FormulaRu
InChI KeyKJTLSVCANCCWHF-UHFFFAOYSA-N
SMILES[Ru]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to dark grey
FormPowder
Total Metal Impurities≤0.1% (≤1000ppm)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only