Nickel(II) oxide, 99% (metals basis)
Nickel(II) oxide, 99% (metals basis)
Nickel(II) oxide, 99% (metals basis)
Nickel(II) oxide, 99% (metals basis)
Thermo Scientific Chemicals

Nickel(II) oxide, 99% (metals basis)

CAS: 1313-99-1 | NiO | 74.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 012359.18
also known as 012359-18
Price (USD)
54.30
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
54.30
Each
Chemical Identifiers
CAS1313-99-1
IUPAC Nameoxonickel
Molecular FormulaNiO
InChI KeyGNRSAWUEBMWBQH-UHFFFAOYSA-N
SMILESO=[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤ 1%
Particle Size-325 Mesh
FormPowder
Appearance (Color)Green
It is used in nickel-iron battery (Edison Battery) and fuel cells. It is a precursor for many nickel salts and nickel alloys including nickel-steel allys, for example invar and elinvar. It is used in electrochromic devices, frits, ferrites, and porcelain glazes. It also serves as a hydrogenation catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only