Aluminum nitrate nonahydrate, 98%
Aluminum nitrate nonahydrate, 98%
Aluminum nitrate nonahydrate, 98%
Thermo Scientific Chemicals

Aluminum nitrate nonahydrate, 98%

CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
5 kg
Catalog number 012360.A1
Price (USD)
140.00
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
140.00
Each
Chemical Identifiers
CAS7784-27-2
IUPAC Namealuminium(3+) nonahydrate trinitrate
Molecular FormulaAlH18N3O18
InChI KeySWCIQHXIXUMHKA-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.5% min
It finds use in tanning leather, antiperspirant, corrosion protection, extraction of uranium, nitrating agent. It is an effective dopant for producing highly conductive thin film by utilizing the sol gel method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only