Ammonium chloride, 99.5% min
Ammonium chloride, 99.5% min
Ammonium chloride, 99.5% min
Thermo Scientific Chemicals

Ammonium chloride, 99.5% min

CAS: 12125-02-9 | ClH4N | 53.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 012361.A3
also known as 012361-A3
Price (USD)
106.65
Online Exclusive
118.00
Save 11.35 (10%)
Each
Quantity:
2 kg
Request bulk or custom format
Price (USD)
106.65
Online Exclusive
118.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS12125-02-9
IUPAC Nameamine hydrochloride
Molecular FormulaClH4N
InChI KeyNLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILESN.Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormGranular
Assay (unspecified)99.5% min
Appearance (Color)White to pale yellow
It act as a buffer, as a flux in galvanizing steel sheet. It is used in batteries, to clean soldering irons, analytical lab reagent, in washing powders, electroplating, lustering cotton.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only