Triethylamine, 99+%
Triethylamine, 99+%
Triethylamine, 99+%
Thermo Scientific Chemicals

Triethylamine, 99+%

CAS: 121-44-8 | C6H15N | 101.193 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
3 x 1 kg
Catalog number 012391.D5
also known as 012391-D5
Price (USD)
162.65
Online Exclusive
181.00
Save 18.35 (10%)
Each
Quantity:
3 x 1 kg
Request bulk or custom format
Price (USD)
162.65
Online Exclusive
181.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS121-44-8
IUPAC Nametriethylamine
Molecular FormulaC6H15N
InChI KeyZMANZCXQSJIPKH-UHFFFAOYSA-N
SMILESCCN(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)≤15 [EU & US]
Impurity contentEthylamine ≤0.1% [US]
Assay (GC)≥99.0% [EU & US]
FormLiquid [EU & US]
Impurity contentDiethylamine ≤0.1% [US]
View more
Triethylamine is a base used to prepare esters and amides from acyl chlorides as well as in the synthesis of quaternary ammonium compounds. It acts as a catalyst in the formation of urethane foams and epoxy resins, dehydrohalogeantion reactions, acid neutralizers for condensation reactions and Swern oxidations. It finds application in reverse phase high-performance liquid chromatography (HPLC) as a mobile-phase modifier. It is also used as an accelerator activator for rubber, as a propellant, as a corrosion inhibitor, as a curing and hardening agent for polymers and for the desalination of seawater. Furthermore, it is used in the automotive casting industry and the textile industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only