Samarium foil, 0.1mm (0.004in) thick, 99.9% (REO)
Samarium foil, 0.1mm (0.004in) thick, 99.9% (REO)
Samarium foil, 0.1mm (0.004in) thick, 99.9% (REO)
Thermo Scientific Chemicals

Samarium foil, 0.1mm (0.004in) thick, 99.9% (REO)

CAS: 7440-19-9 | Sm | 150.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 012421.FF
Price (USD)
-
Quantity:
25 x 25 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-19-9
IUPAC Namesamarium
Molecular FormulaSm
InChI KeyKZUNJOHGWZRPMI-UHFFFAOYSA-N
SMILES[Sm]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only