Osmium powder, -200 mesh, 99.8% (metals basis)
Osmium powder, -200 mesh, 99.8% (metals basis)
Osmium powder, -200 mesh, 99.8% (metals basis)
Thermo Scientific Chemicals

Osmium powder, -200 mesh, 99.8% (metals basis)

CAS: 7440-04-2 | Os | 190.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 012446.03
also known as 012446-03
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS7440-04-2
IUPAC Nameosmium
Molecular FormulaOs
InChI KeySYQBFIAQOQZEGI-UHFFFAOYSA-N
SMILES[Os]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
Total Metal Impurities≤0.2%
Particle Size-200 mesh
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only