Gold flake/sphere, APS 1-15 micron, 99.95% (metals basis)
Gold flake/sphere, APS 1-15 micron, 99.95% (metals basis)
Gold flake/sphere, APS 1-15 micron, 99.95% (metals basis)
Thermo Scientific Chemicals

Gold flake/sphere, APS 1-15 micron, 99.95% (metals basis)

CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 012453.03
also known as 012453-03
Price (USD)
322.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
322.00
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-57-5
ChEBICHEBI:30050
Chemical Name or MaterialGold flake/sphere
EINECS Number231-165-9
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only