Copper(II) chloride dihydrate, ACS, 99+%
Copper(II) chloride dihydrate, ACS, 99+%
Copper(II) chloride dihydrate, ACS, 99+%
Copper(II) chloride dihydrate, ACS, 99+%
Thermo Scientific Chemicals

Copper(II) chloride dihydrate, ACS, 99+%

CAS: 10125-13-0 | Cl2CuH4O2 | 170.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 012458.A3
also known as 012458-A3
Price (USD)
436.65
Special offer
Online exclusive
Ends: 30-Jun-2026
513.00
Save 76.35 (15%)
Each
Quantity:
2 kg
Request bulk or custom format
Price (USD)
436.65
Special offer
Online exclusive
Ends: 30-Jun-2026
513.00
Save 76.35 (15%)
Each
Chemical Identifiers
CAS10125-13-0
IUPAC Namecopper(2+) dihydrate dichloride
Molecular FormulaCl2CuH4O2
InChI KeyMPTQRFCYZCXJFQ-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Cu++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥99.0% CuCl₂•2H₂O
Impurity contentCa: ≤0.005%
Impurity contentFe: ≤0.005%
Impurity contentK: ≤0.01%
Impurity contentNa: ≤0.02%
View more

Polystyrene, atactic is used as plastics, include protective packaging, containers, lids, bottles, trays, tumblers, and disposable cutlery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only