Bismuth Tin eutectic lump, 99.95% (metals basis)
Thermo Scientific Chemicals

Bismuth Tin eutectic lump, 99.95% (metals basis)

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 012480.14
also known as 012480-14
Price (USD)
48.20
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
48.20
Each
Specifications
Assay Percent Notes(metals basis)
Avg. Mol. Wt. or Mol. Wt. RangeBi:Sn; 58:42 wt%
Chemical Name or MaterialBismuth Tin eutectic lump
MDL NumberMFCD00143771
Melting Point138°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only