Iron(II) chloride hydrate, Reagent Grade, 99%
Iron(II) chloride hydrate, Reagent Grade, 99%
Iron(II) chloride hydrate, Reagent Grade, 99%
Iron(II) chloride hydrate, Reagent Grade, 99%
Thermo Scientific Chemicals

Iron(II) chloride hydrate, Reagent Grade, 99%

Air Sensitive | CAS: 23838-02-0 | Cl2Fe . x H2O | 126.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
5 kg
Catalog number 012484.14
also known as 012484-14
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.00
Save 7.35 (14%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.00
Save 7.35 (14%)
Each
Chemical Identifiers
CAS23838-02-0
IUPAC Nameλ²-iron(2+) dichloride
Molecular FormulaCl2Fe . x H2O
InChI KeyNMCUIPGRVMDVDB-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Fe++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
It is used as a reducing agent in metallurgy, in pharmaceutical preparations, as a mordant in dyeing and in sewage treatment. Ferrous chloride has a kind of niche application. During the laboratory synthesis of iron complexes, ferrous chloride acts as a reducing flocculating agent in wastewater treatment, particularly for wastes containing chromate. It is the precursor to hydrated iron(III) oxides that are magnetic pigments. Ferrous chloride is employed as a reducing agent in many organic synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only