Chromium(VI) oxide, 99%
Chromium(VI) oxide, 99%
Chromium(VI) oxide, 99%
Thermo Scientific Chemicals

Chromium(VI) oxide, 99%

CAS: 1333-82-0 | CrO3 | 99.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 x 1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012522.30
also known as 012522-30
Price (USD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS1333-82-0
IUPAC Nametrioxochromium
Molecular FormulaCrO3
InChI KeyWGLPBDUCMAPZCE-UHFFFAOYSA-N
SMILESO=[Cr](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to dark red to purple
FormFlakes or prills or crystals or granules
Assay from Supplier's CofA≥98.5 to ≤101.5%
It is widely used in chromium electroplating, and as protective coatings in corrosion/oxidation resistance of metals. It is also employed in CDs and DVDs. It is useful as an oxidant in Jones oxidation in acetic acid or acetone. It is used in the oxidation of primary alcohols into carboxylic acids, and secondary alcohols into ketones. With phosphoric acid, it is used as a stripping agent for anodic coatings of all types and in the production of syntheric rubies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only