Copper(II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%
Copper(II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%
Copper(II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%
Thermo Scientific Chemicals

Copper(II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%

For Use As a Catalyst, Nitrating Agent For Aromatic organosilicon Compounds, Nickel-Plating Baths | CAS: 19004-19-4 | CuN2O6·2.5H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
500 g
2 kg
10 kg
Catalog number 012523.36
also known as 012523-36
Price (USD)
112.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
112.00
Each
Chemical Identifiers
CAS19004-19-4
IUPAC Namedicopper(2+) pentahydrate tetranitrate
Molecular FormulaCu2H10N4O17
InChI KeyIZDJJEMZQZQQQQ-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0-102.0% CuN₂O₆.2.5H₂O
Impurity contentCa: ≤0.005%
Impurity contentK: ≤0.005%
Impurity contentCl: ≤0.002%
Impurity contentFe: ≤0.005%
View more
Catalyst, nitrating agent for aromatic organosilicon compounds, nickel-plating baths, coloring copper black and producing a burnished effect on ironCopper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only