Manganese(III) 2,4-pentanedionate
Manganese(III) 2,4-pentanedionate
Manganese(III) 2,4-pentanedionate
Thermo Scientific Chemicals

Manganese(III) 2,4-pentanedionate

CAS: 14284-89-0 | C15H21MnO6 | 352.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 012530.30
also known as 012530-30
Price (USD)
250.65
Online Exclusive
278.00
Save 27.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
250.65
Online Exclusive
278.00
Save 27.35 (10%)
Each
Specifications
CAS14284-89-0
Chemical Name or MaterialManganese(III) 2,4-pentanedionate
EINECS Number238-188-3
Formula Weight352.24
Health Hazard 1H302+H312+H332-H315-H319-H335
View more
It is mainly used as a solvent in paint and a precursor to vitamin E.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only