Platinum rod, 6.35mm (0.25in) dia, Premion™, 99.99% (metals basis)
Platinum rod, 6.35mm (0.25in) dia, Premion™, 99.99% (metals basis)
Platinum rod, 6.35mm (0.25in) dia, Premion™, 99.99% (metals basis)
Thermo Scientific Chemicals

Platinum rod, 6.35mm (0.25in) dia, Premion™, 99.99% (metals basis)

CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantity
012556.HF
also known as 012556-HF
10 mm
Catalog number 012556.HF
also known as 012556-HF
Price (USD)
-
Quantity:
10 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
FormRod, approx. 6.35 mm dia.
Appearance (Color)Metallic grey
Comment99.99% Platinum (metals basis): Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only