Tin plate 3.2mm (0.13in) thick, 99.85% (metals basis)
Tin plate 3.2mm (0.13in) thick, 99.85% (metals basis)
Tin plate 3.2mm (0.13in) thick, 99.85% (metals basis)
Thermo Scientific Chemicals

Tin plate 3.2mm (0.13in) thick, 99.85% (metals basis)

CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
100 x 500 mm
Catalog number 012602.GH
also known as 012602-GH
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-31-5
ChEBICHEBI:32990
Chemical Name or MaterialTin plate
EINECS Number231-141-8
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only