Potassium bromide, ACS, 99% min
Potassium bromide, ACS, 99% min
Potassium bromide, ACS, 99% min
Thermo Scientific Chemicals

Potassium bromide, ACS, 99% min

For Manufacturing of Photographic Papers, Process Engraving and Lithography, Spectroscopy | CAS: 7758-02-3 | BrK | 119.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
5 kg
Catalog number 012612.A7
also known as 012612-A7
Price (USD)
402.65
Special offer
Online exclusive
Ends: 30-Jun-2026
474.00
Save 71.35 (15%)
Each
Quantity:
5 kg
Request bulk or custom format
Price (USD)
402.65
Special offer
Online exclusive
Ends: 30-Jun-2026
474.00
Save 71.35 (15%)
Each
Chemical Identifiers
CAS7758-02-3
IUPAC Namepotassium bromide
Molecular FormulaBrK
InChI KeyIOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES[K+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or crystalline powder or solid chunks
Assay (Argentometric Titration)≥99.0%
Barium content≤0.002%
Bromate content≤0.001%
View more
Used in manufacturing of photographic papers, engraving and lithography, in spectroscopy, and as an analytical lab reagent. Potassium bromide is applied as a veterinary drug, as an antiepileptic medication for dogs. It is utilized for the bromination of alkenes using cerium(IV) ammonium nitrate. It plays an essential role with diacetoxy iodobenzene for the efficient bromination of alkenes and alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only