Ammonium aquopentachlororhodate(III), Rh 30% min
Ammonium aquopentachlororhodate(III), Rh 30% min
Ammonium aquopentachlororhodate(III), Rh 30% min
Thermo Scientific Chemicals

Ammonium aquopentachlororhodate(III), Rh 30% min

CAS: 63771-33-5 | Cl5H10N2ORh | 334.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 012645.03
also known as 012645-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS63771-33-5
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Rh 30% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only