Potassium osmium(VI) oxide dihydrate, 98+%
Potassium osmium(VI) oxide dihydrate, 98+%
Potassium osmium(VI) oxide dihydrate, 98+%
Thermo Scientific Chemicals

Potassium osmium(VI) oxide dihydrate, 98+%

CAS: 10022-66-9 | H4K2O6Os | 368.453 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
Catalog number 012647.01
also known as 012647-01
Price (USD)
69.65
Special offer
Online exclusive
Ends: 28-Dec-2026
81.80
Save 12.15
Each
Quantity:
0.25 g
Request bulk or custom format
Price (USD)
69.65
Special offer
Online exclusive
Ends: 28-Dec-2026
81.80
Save 12.15
Each
Chemical Identifiers
CAS10022-66-9
IUPAC Namedipotassium dioxoosmiumbis(olate) dihydrate
Molecular FormulaH4K2O6Os
InChI KeyDGODWNOPHMXOTR-UHFFFAOYSA-N
SMILESO.O.[K+].[K+].[O-][Os]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥98.0%
Appearance (Color)Purple
FormPowder
Potassium osmium(VI) oxide dihydrate acts as a catalyst for the oxidative cleavage of olefins to ketones and carboxylic acids. It is employed in an intramolecular amidohydroxylation of carbamoyloxytethered olefins providing hydroxy oxazolidinones. It is also used as a reagent in the synthesis of amphidinolide B, which shows potent antitumor activity against human solid and blood tumor cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only