Lithium sulfide, 99.9% (metals basis) - FAQs
Lithium sulfide, 99.9% (metals basis) - FAQs
View additional product information for Lithium sulfide, 99.9% (metals basis) - FAQs (012839.30, 012839.09, 012839.18, 012839.04)
1 product FAQs found
How should Lithium sulfide, 99.9% (metals basis) (Cat. No. 012839.xx) be stored and handled?
Lithium sulfide releases flammable gas when in contact with water and should be kept away from water, moist air, or acids. Lithium sulfide should be stored and handled under a dry, inert gas atmosphere. Store at ambient temperatures, typically 20-25 degrees C.