Aluminum sulfide, 99+%, (metals basis)
Aluminum sulfide, 99+%, (metals basis)
Aluminum sulfide, 99+%, (metals basis)
Thermo Scientific Chemicals

Aluminum sulfide, 99+%, (metals basis)

CAS: 1302-81-4 | Al2S3 | 150.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012851.06
also known as 012851-06
Price (USD)
213.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
213.00
Each
Chemical Identifiers
CAS1302-81-4
IUPAC Namedialuminium(3+) trisulfanediide
Molecular FormulaAl2S3
InChI KeyCOOGPNLGKIHLSK-UHFFFAOYSA-N
SMILES[Al+3].[Al+3].[S--].[S--].[S--]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only