Strontium sulfate, Puratronic™, 99.9965% (metals basis)
Strontium sulfate, Puratronic™, 99.9965% (metals basis)
Strontium sulfate, Puratronic™, 99.9965% (metals basis)
Thermo Scientific Chemicals

Strontium sulfate, Puratronic™, 99.9965% (metals basis)

CAS: 7759-02-6 | O4SSr | 183.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 012870.06
also known as 012870-06
Price (USD)
74.65
Online exclusive
82.70
Save 8.05
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
74.65
Online exclusive
82.70
Save 8.05
Each
Chemical Identifiers
CAS7759-02-6
IUPAC Namestrontium(2+) sulfate
Molecular FormulaO4SSr
InChI KeyUBXAKNTVXQMEAG-UHFFFAOYSA-L
SMILES[Sr++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0035% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only