Cesium fluoride, 99% (metals basis)
Cesium fluoride, 99% (metals basis)
Cesium fluoride, 99% (metals basis)
Thermo Scientific Chemicals

Cesium fluoride, 99% (metals basis)

CAS: 13400-13-0 | CsF | 151.90 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 012885.22
also known as 012885-22
Price (USD)
168.65
Online Exclusive
187.00
Save 18.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
168.65
Online Exclusive
187.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS13400-13-0
IUPAC Namecaesium(1+) fluoride
Molecular FormulaCsF
InChI KeyXJHCXCQVJFPJIK-UHFFFAOYSA-M
SMILES[F-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Used for desilylation reactions in organic synthesis. Useful base in organic chemistry, due to its low nucleophilicity. Owing to its high ionic character, it is a very reactive source of fluoride ion in organic synthesis. Reagent of choice when reactive naked fluoride is required. Effective catalyst in promoting Knoevenegal condensation. Single crystals of CsF are useful as the windows of cells for infrared spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only