Rubidium bromide, 99.8% (metals basis)
Rubidium bromide, 99.8% (metals basis)
Rubidium bromide, 99.8% (metals basis)
Thermo Scientific Chemicals

Rubidium bromide, 99.8% (metals basis)

CAS: 7789-39-1 | BrRb | 165.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 012891.06
also known as 012891-06
Price (USD)
45.65
Special offer
Online exclusive
Ends: 30-Jun-2026
53.10
Save 7.45 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
45.65
Special offer
Online exclusive
Ends: 30-Jun-2026
53.10
Save 7.45 (14%)
Each
Chemical Identifiers
CAS7789-39-1
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Rubidium bromide is used in infrared optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only