Gadolinium powder, -40 mesh, 99% (REO)
Gadolinium powder, -40 mesh, 99% (REO)
Gadolinium powder, -40 mesh, 99% (REO)
Thermo Scientific Chemicals

Gadolinium powder, -40 mesh, 99% (REO)

CAS: 7440-54-2 | Gd | 157.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 012896.04
also known as 012896-04
Price (USD)
52.80
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
52.80
Each
Chemical Identifiers
CAS7440-54-2
IUPAC Namegadolinium
Molecular FormulaGd
InChI KeyUIWYJDYFSGRHKR-UHFFFAOYSA-N
SMILES[Gd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only